Please login to your online account to display your discounted pricing

OHAUS™ BALANCE 2200G X 0.01G PROMO

The Explorer risen to the next level of ingenuity; expanding on its modern features and design to offer unmatched functionality. Note: Promo offer expires Sept. 30, 2016.

$2,853.91 - $9,098.33

Specifications

Amperage 0.6A
Automation Interface Touch screen
Depth (Metric) 39.3cm
Depth (English) 15.5 in.
Connection Type RS232, USB, Optional Ethernet or 2nd RS232
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Type Model Readability Price Quantity    

02112233PM

 
ohaus™
30061976PM
120g Analytical/Precision Balance EX124/AD 0.0001g Each for $7,210.49

02112234PM

 
ohaus™
30061977PM
220g Analytical/Precision Balance EX224/AD 0.0001g Each for $7,791.37

02112235PM

 
ohaus™
30061978PM
320g Analytical/Precision Balance EX324/AD 0.0001g Each for $8,662.68

02112236PM

 
ohaus™
30061997PM
120g Analytical/Precision Balance (NTEP Model) EX124N/AD 0.0001g Each for $7,646.14

02112237PM

 
ohaus™
30061998PM
220g Analytical/Precision Balance (NTEP Model) EX224N/AD 0.0001g Each for $8,227.02

02112238PM

 
ohaus™
30061999PM
320g Analytical/Precision Balance (NTEP Model) EX324N/AD 0.0001g Each for $9,098.33

02112239PM

 
ohaus™
30061979PM
220g Analytical/Precision Balance EX223/E 0.001g Each for $2,853.91

02112240PM

 
ohaus™
30061980PM
420g Analytical/Precision Balance EX423/E 0.001g Each for $3,580.00

02112241PM

 
ohaus™
30061981PM
2200g Analytical/Precision Balance EX2202/E 0.01g Each for $3,144.35

02112243PM

 
ohaus™
30061983PM
6200g Analytical/Precision Balance EX6202/E 0.01g Each for $4,160.88

02112242PM

 
ohaus™
30061982PM
4200g Analytical/Precision Balance EX4202/E 0.01g Each for $3,652.61

02112244PM

 
ohaus™
30061984PM
6200g Analytical/Precision Balance EX6201/E 0.1g Each for $3,434.79
Description & Specifications

Specifications

Amperage 0.6A
Automation Interface Touch screen
Depth (Metric) 39.3cm
Depth (English) 15.5 in.
Connection Type RS232, USB, Optional Ethernet or 2nd RS232
Width (Metric) 23cm
Width (English) 9.1 in.
Display Type Color Graphic Display
Electrical Requirements 100/240V 50/60Hz
Format Gram, milligram, kilogram, carat, ounce, troy ounce, pound, pennyweight, grain, Newton, momme, mesghal, Hong Kong tael, Singapore tael, Taiwan tael, Tical, Tola, Baht, custom unit 1, custom unit 2, custom unit 3
Hertz 50/60Hz
Material ABS Plastic, Stainless Steel, Glass
Sensitivity Drift 1.5ppm/°C
Temperature Range (Metric) 10° to 30°C
Temperature Range (English) 50° to 86°F
Voltage 100/240V

  • Large color touch-screen display
  • Icon-driven application software that features 14 unique application modes
  • 3 level adjustable angle display
  • Newly optimized linearity and repeatability specifications
  • Enhanced vibration filtering for better balance stability
  • All models come standard with AutoCal™ automatic internal calibration
  • 3 x 3 filtering setup ensures fast and stable weighing results
  • The base and display separate for maximum flexibility in anywork area
  • Draftshield models isolate the weighing chamber to enhance stability and speed results while offering maximum accessibility to the weighing chamber through top-mounted side doors or through the versatile top door that flips or slides open
  • Hands-free entry is available with optional automated side doors
  • Easy access communication ports (USB, RS232) plus 3rd communication port slot (Ethernet or 2nd RS232)
  • GLP and GMP including date and time, Direct Data Transfer
  • Metal base, ABS top housing, stainless steel pan, glass draftshield with flip-top door, replaceable in-use cover
  • 3-door draftshield with removable panels on 0.1mg and 1 mg models with draftshield light
  • Modular display design with adjustable angle (attach to base, wall or optional tower)
  • AutoCal automatic internal calibration
  • Level assist feature with illuminated level indicator
  • Selectable environmental and auto-print settings
  • Integral weigh belowhook
  • Protective in-use cover
  • Software lock switch
  • High resolution 14.5cm (diagonal) full colour VGA touch display including QWERTY and numeric key pads plus weighing capacity indication
Applications
Weighing (17 units + custom units), parts counting, percent weighing, animal/dynamic weighing, checkweighing, filling,totalization, formulation, differential weighing, density determination, peak hold, ingredient costing, pipette adjustment, gross/net/tare weighing

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.