Please login to your online account to display your discounted pricing

ExpiCHO-S™ Cells for ExpiCHO Expression System

Maintained in suspension culture, these cells have minimal tendency to clump, and grow to high density in ExpiCHO™ Expression Medium

$1,090.00 - $4,025.00

For Use With (Application) ExpiCHO Expression System
View More Specs
Products
Catalog Number Mfr. No. Packaging Price Quantity    

A29127

 
gibco
LSA29127
1 vial Each for $1,090.00

A29132

 
gibco
LSA29132
6-vial pack Each for $4,025.00
Description & Specifications

Specifications

For Use With (Application) ExpiCHO Expression System
Storage Requirements Store in liquid Nitrogen

  • Allow yields of up to 3g of protein per L of transfected culture as part of the ExpiCHO expression system
  • Enable rapid, high-yield transient protein production, allowing labs developing protein biotherapeutics to work with CHO-expressed proteins from start to finish in the drug development process
  • Adapted to high-density suspension growth in ExpiCHO Expression Medium
  • Exhibit rapid doubling times of approximately 17 hours
  • Enable consistent transient expression performance for over 30 passages after thaw
  • Maintain high cell viability and productivity for greater than 10 days post-transfection as part of the ExpiCHO expression system