missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.
Supplier: Applied Biosystems™ 7820510ML
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
| Shipped on dry ice. Store at -20°C. | |
| PCR Products | |
| 5000 Reactions | |
| Recombinant | |
| Genetic Diseases |
| ExoSAP-IT™ PCR Product Clean-up Reagent, 5000 Reactions, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification | |
| PCR Purification | |
| ExoSAP-IT™ | |
| 10 mL | |
| Dry Ice |