Please login to your online account to display your discounted pricing
Affymetrix™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications.
$60.90 - $5,298.30
Specifications
Storage Requirements −20°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
501124988
affymetrix™
750011ML
500 reactions    Each for $651.05
501124990
affymetrix™
7500140UL
20 reactions    Each for $60.90
501124989
affymetrix™
750014X1ML
2,000 reactions    Each for $2,341.75
501129341
affymetrix™
7500110ML
5,000 reactions    Each for $5,298.30
Description & Specifications
Specifications
Storage Requirements −20°C
  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows