Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications.
Manufacturer:  Applied Biosystems™ LS750014X1ML
 View more versions of this product
Catalog No. 501124989
Description
Description
  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
Specifications
Specifications
2,000 reactions   
−20°C
Documents