Please login to your online account to display your discounted pricing

Exo1, Tocris

Inhibits Golgi-ER traffic; blocks exocytosis

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Exo1
Assay Percent Range >99%
Molecular Formula C15H12FNO3
Formula Weight 273.26
Synonym 2-[(4-Fluorobenzoyl)amino]-benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

185050

 
r & d systems
RDS185050
50mg Each for $598.33

185010

 
r & d systems
RDS185010
10mg Each for $142.08
Description & Specifications

Specifications

Formula Weight 273.26
Quantity 50mg
Chemical Name or Material Exo1
Assay Percent Range >99%
Molecular Formula C15H12FNO3
CAS 461681-88-9
Synonym 2-[(4-Fluorobenzoyl)amino]-benzoic acid methyl ester
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature