Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Exo1 Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Exo1 Ab-4, Mouse Monoclonal Antibody.

$303.06 - $1,256.61

Specifications

Antigen Exo1 Ab-4
Clone 266
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1534P

 
thermo scientific™ lab vision™
MS1534P
200μg/mL 1mL Each for $1,171.16

MS1534P0

 
thermo scientific™ lab vision™
MS1534P0
200μg/mL 100µL Each for $303.06

MS1534P1

 
thermo scientific™ lab vision™
MS1534P1
200μg/mL 500µL Each for $750.88

MS1534P1ABX

 
thermo scientific™ lab vision™
MS1534P1ABX
1mg/mL 100µL Each for $853.59

MS1534PABX

 
thermo scientific™ lab vision™
MS1534PABX
1mg/mL 200µL Each for $1,256.61
Description & Specifications

Specifications

Antigen Exo1 Ab-4
Clone 266
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant full length mouse Exo1 protein
Isotype IgG2a kappa

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Exo1 Ab-4, Mouse Monoclonal Antibody.

The 5' to 3' double-strand DNA exonuclease (Exo1) is coded by the EXO1 gene. It was identified in Saccharomyces cerevisiae as a gene encoding an exonuclease that interacts with meiosis-specific MutS homologue (MSH2) and functions in mismatch repair and genetic recombination.

Host Species: Mouse

Clone: 266

Isotype: IgG1/κ

Species Reactivity: Human and Mouse. Others not known.

Immunogen: Recombinant full-length mouse Exo1 protein

Molecular Weight: ∽92 kDa

Positive Control: MCF-7 and MAD109

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO