Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Exo1 Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Exo1 Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1534P1ABX

 View more versions of this product

Catalog No. MS1534P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Exo1 Ab-4
Applications Western Blot
Classification Monoclonal
Clone 266
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Murine
Immunogen Recombinant full length mouse Exo1 protein
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Murine

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Exo1 Ab-4, Mouse Monoclonal Antibody.

The 5' to 3' double-strand DNA exonuclease (Exo1) is coded by the EXO1 gene. It was identified in Saccharomyces cerevisiae as a gene encoding an exonuclease that interacts with meiosis-specific MutS homologue (MSH2) and functions in mismatch repair and genetic recombination.

Host Species: Mouse

Clone: 266

Isotype: IgG1/κ

Species Reactivity: Human and Mouse. Others not known.

Immunogen: Recombinant full-length mouse Exo1 protein

Molecular Weight: ∽92 kDa

Positive Control: MCF-7 and MAD109

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO