Please login to your online account to display your discounted pricing

exo-IWR 1, Tocris™

Negative control for endo-IWR 1

$175.83 - $734.99

Chemical Identifiers

Chemical Name or Material exo-IWR 1
Assay Percent Range >99%
Molecular Formula C25H19N3O3
Formula Weight 409.44
Synonym 4-[(3aR,4R,7S,7aS-rel)-1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl]-N-8-quinolinylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394710

 
r & d systems
RDS394710
10mg Each for $175.83

394750

 
r & d systems
RDS394750
50mg Each for $734.99
Description & Specifications

Specifications

Chemical Name or Material exo-IWR 1
Quantity 10mg
CAS 1127442-87-8
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C25H19N3O3
Formula Weight 409.44
Synonym 4-[(3aR,4R,7S,7aS-rel)-1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl]-N-8-quinolinylbenzamide
Safety and Handling

Store at Room Temperature