Please login to your online account to display your discounted pricing

Alfa Aesar™ exo-6-Hydroxytropinone, 98+%

Chemical Identifiers

Chemical Name or Material exo-6-Hydroxytropinone
CAS 5932-53-6
Molecular Formula C8H13NO2
Formula Weight 155.19
MDL Number MFCD00135648
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5600706

 
alfa aesar™
H5600706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.19
Chemical Name or Material exo-6-Hydroxytropinone
Assay ≥98%
Molecular Formula C8H13NO2
MDL Number MFCD00135648
CAS 5932-53-6
Melting Point 120° to 121°C
Quantity 5g