missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ exo-6-Hydroxytropinone, 98+%

Manufacturer:  Alfa Aesar™ H5600706

 View more versions of this product

Catalog No. AAH5600706



Specifications

Specifications

exo-6-Hydroxytropinone
5g
C8H13NO2
1r,5r-6-hydroxy-8-methyl-8-azabicyclo 3.2.1 octan-3-one
CN1C2CC(C1CC(=O)C2)O
155.197
155.19
120°C to 121°C
5932-53-6
MFCD00135648
UOHSTKWPZWFYTF-LSBSRIOGSA-N
(1R,5R)-6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one
51346151
≥98%

Chemical Identifiers

5932-53-6
155.197
UOHSTKWPZWFYTF-LSBSRIOGSA-N
51346151
CN1C2CC(C1CC(=O)C2)O
C8H13NO2
MFCD00135648
1r,5r-6-hydroxy-8-methyl-8-azabicyclo 3.2.1 octan-3-one
(1R,5R)-6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one
Safety and Handling

Safety and Handling

EINECSNumber : 227-677-7

TSCA : No

Recommended Storage : Ambient temperatures

Documents