missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride, TCI America™

Manufacturer:  TCI America E0392500G

 View more versions of this product

Catalog No. E0392500G



Specifications

Specifications

exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride
500g
C8H6O4
exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
C1=CC2C3C(C1O2)C(=O)OC3=O
124671625
Crystal-Powder at 20°C
White
6118-51-0
MFCD00151506
QQYNRBAAQFZCLF-ZXXMMSQZSA-N
166.132
166.13
≥98.0% (T)

Chemical Identifiers

6118-51-0
166.132
QQYNRBAAQFZCLF-ZXXMMSQZSA-N
124671625
C8H6O4
MFCD00151506
exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
C1=CC2C3C(C1O2)C(=O)OC3=O
Safety and Handling

Safety and Handling

RTECSNumber : KC9650000

TSCA : Yes

Documents