Welch™ Exhaust Filters for Welch™ DuoSeal™, Chemstar™ and Direct-Drive Pumps
Prevent damage to your pump by choosing to use an exhaust fulter from Welch for Duo Seal and Direct-Drive model pumps.
Manufacturer: Welch™ 1417B
Description
Easily replaceable filter element captures oil mist from the exhaust port of the pump and reduces pump noise.
Specifications
Welch Pump Models 1397, 1374 | |
Directional Outlet with Exhaust Tubing Connector | |
Exhaust Filter |