Please login to your online account to display your discounted pricing

Alfa Aesar™ Exendin 4

For cellular and molecular biology applications

$306.31 - $306.31

Specifications

Product Type Exendin 4
Molecular Weight 4186.56
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66726EXD

 
alfa aesar™
J66726EXD
0.5mg Each for $306.31

AAJ66726MCR

 
alfa aesar™
J66726MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Exendin 4
Molecular Weight 4186.56
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC