Please login to your online account to display your discounted pricing

Alfa Aesar™ Exendin 4


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66726EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66726EXD

  • / Each
 

Description & Specifications

Specifications

Product Type Exendin 4
Format Solid
Molecular Weight 4186.56
Quantity 0.5mg
Purification Method HPLC
Storage Requirements -30° to -10°C
Regulatory Status RUO