Please login to your online account to display your discounted pricing

Exemestane, Tocris™

Steroidal aromatase (CYP19) inhibitor

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material Exemestane
Assay Percent Range >99%
Molecular Formula C20H24O2
Formula Weight 296.4
Synonym 6-Methyleneandrosta-1, 4-diene-3, 17-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

375910

 
r & d systems
RDS375910
10mg Each for $205.70

375950

 
r & d systems
RDS375950
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 296.4
Quantity 10mg
Chemical Name or Material Exemestane
CAS 107868-30-4
Synonym 6-Methyleneandrosta-1, 4-diene-3, 17-dione
Assay Percent Range >99%
Molecular Formula C20H24O2
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C