Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Excitatory Amino Acid Transporter 2 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Excitatory Amino Acid Transporter 2 Ab-1, Mouse Monoclonal Antibody.

$205.88 - $883.12

Specifications

Antigen Excitatory Amino Acid Transporter 2 Ab-1
Clone 1H8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1750R7

 
thermo scientific™ lab vision™
MS1750R7
7mL Each for $511.04

MS1750S

 
thermo scientific™ lab vision™
MS1750S
1mL Each for $883.12

MS1750S0

 
thermo scientific™ lab vision™
MS1750S0
100µL Each for $205.88

MS1750S1

 
thermo scientific™ lab vision™
MS1750S1
500µL Each for $660.04
Description & Specifications

Specifications

Antigen Excitatory Amino Acid Transporter 2 Ab-1
Clone 1H8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Recombinant protein corresponding to C-terminal cytoplasmic domain of human EAAT2
Isotype IgG2a

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Excitatory Amino Acid Transporter 2 Ab-1, Mouse Monoclonal Antibody.

Human excitatory amino acid transporters regulate neurotransmitter concentrations at the excitatory glutamatertic synapses of the mammalian central nervous system. They reduce extracellular glutamate concentration, modulating synaptic signaling, which helps reduce the risk of excitotoxicity. Expression of EATT2 is restricted to motor cortex area of the brain. EATT1 blocks the transport mediated by EATT2. EATT2 shares homology to L-glutamate transporter (GLT-1) from brain tissue.

Host Species: Mouse

Clone: 1H8

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: C-terminal cytoplasmic domain

Immunogen: Recombinant protein corresponding to C-terminal cytoplasmic domain of human EAAT2

Positive Control: Brain

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO