Please login to your online account to display your discounted pricing
Excelta™ High Precision Loupes
Two-element optical glass construction
$177.33 - $177.33
Specifications
Product Type High Precision Eye Loupe
For Use With (Application) To admit light from all directions and a wide range of view
Material Optical Glass
View More Specs
Products
Catalog Number Mfr. No. Magnification Price Quantity    
17467627
excelta corporation
405
5x Each for $177.33
Description & Specifications
Specifications
Product Type High Precision Eye Loupe
For Use With (Application) To admit light from all directions and a wide range of view
Material Optical Glass
  • Bright, sharp focus and a wide range of view
  • 360 degree clear acrylic base to admit light from all directions