Please login to your online account to display your discounted pricing
Excelta™ Five Star Porta-Wand Elite Wafer Pickup System
Vacuum handling tools, kits and accessories
$8,253.79 - $8,253.79
Specifications
Product Type Vacuum Tool Kit
For Use With (Application) Ideal for handling wafers, disk media, flat panels, any item that has a flat hard surface
For Use With (Equipment) Press fit tips, adapters and vacuum hoses
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
17468519
excelta corporation
PV500W6
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 6 in. wafer tip Each for $8,253.79
17468521
excelta corporation
PV500W12
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 12 in. wafer tip CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17468518
excelta corporation
PV500W4
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 4 in. wafer tip Each for $8,253.79
17468520
excelta corporation
PV500W8
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 8 in. wafer tip Each for $8,253.79
Description & Specifications
Specifications
Product Type Vacuum Tool Kit
For Use With (Application) Ideal for handling wafers, disk media, flat panels, any item that has a flat hard surface
For Use With (Equipment) Press fit tips, adapters and vacuum hoses
Vacuum pick-up hand-piece
  • Stainless steel wafer wand
  • Lightweight
  • With push-button
  • Can be used with press-fit tips, adapters and vacuum hoses
  • For 0.13 in. (3.2mm) I.D. hose
Vacuum tool kit contents:
  • Vacuum pick-up hand-piece
  • Detachable 9.6V NiMH rechargeable battery pack
  • In-stand charger
  • Rear exhaust filter
  • Power-on indicator
  • Push button with on/off and lock-to-on
  • Intake air filter
  • Choice of wafer tip