missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Excelta™ Electronic Style Tweezers With Anti-Crush Feature

For general assembly and light magnification

$160.82 - $160.82

Specifications

Material Stainless Steel, Neverrust™
Product Type Tweezers, Electronic
Quantity (No. per Case) Each
Height (English) Body 0.015 in.
Height (Metric) Body 0.038 cm
View More Specs

Products 1
Catalog Number Mfr. No. Length (English) Tip Style Thickness (English) Tip Width (English) Tip Price Quantity  
Catalog Number Mfr. No. Length (English) Tip Style Thickness (English) Tip Width (English) Tip Price Quantity  
17467270 View Documents Excelta™
52ASA
4.75 in. Flat, Round 0.007 in. 0.059 in. Each for $160.82
 
Description

Description

  • High Precision
  • NEVERUST™ stainless steel

Specifications

Specifications

Stainless Steel, Neverrust™
Each
0.038 cm
0.81 cm
Tweezers, Electronic
0.015 in.
0.32 in.