missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Excelta™ Carbofib Tip Tweezers: Replacement Tips

Screw-on tips in assorted point styles

Supplier:  Excelta™ 759RTX

 View more versions of this product

Catalog No. 17467358



Description

Description

  • Conductive tips with surface resistivity: 103 ohm cm
  • Attach to stainless-steel tweezer body

Specifications

Specifications

Curved
Each
0.1 cm
Replacement Tip Kit for 759-RT tweezers
2 tips and 4 screws
SDS
Documents

Documents

Promotions

Promotions