missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Excellence XSR Precision Balances, Standard Weigh Pan

XSR precision balances offer a winning combination of reliable performance and truly ergonomic operation. Even under the harshest of weighing conditions, you can rely on these robust balances to help you meet your productivity targets.

Manufacturer:  Mettler Toledo™ 30317501

 View more versions of this product

Catalog No. 01-919-051



Description

Description

XSR precision balances offer a winning combination of reliable performance and truly ergonomic operation. Even under the harshest of weighing conditions, you can rely on these robust balances to help you meet your productivity targets. Reliable performance, ergonomic weighing processes and easy data transfer ensure you get the accurate results you want quickly and efficiently. XSR balances are easy and comfortable to use, protecting your health during your daily weighing tasks.

  • 4.3 in. color capacitive touchscreen terminal, glove compatible
  • Storage of weighing methods
  • Integrated results notepad
  • Multiple interfaces for easy connectivity and data export: 4 x USB, 1 LAN, 1 RS232
  • Easy connection of peripheral devices: printers, PC, barcode readers, footswitches and sensors
  • Export of process and results data to XML or Excel
  • Customizable tolerance profiles, with pre-defined quality and weighing settings
  • MinWeigh warning function
  • Balance ready StatusLight™
  • LevelControl: level warning with graphical leveling guide
  • Temperature and time programmable automatic adjustment (proFACT)
  • Password protected configuration
  • Integrated overload protection
  • Chemically resistant materials
  • Designed for easy cleaning 
Specifications

Specifications

4.1 kg
FDA 21 CFRpart 11
XSR4001S
USB, LAN, RS232 compatible
0.1 g
Color Graphic Display
Documents
Promotions

Promotions