Please login to your online account to display your discounted pricing
EVOS™ Vessel Holders
For flasks, trays and petri dishes
$173.06 - $217.77
Specifications
Product Type Vessel Holder
View More Specs
Products
Catalog Number Mfr. No. Device Holds For Use With (Equipment) Price Quantity    
12-563-521
evos™
AMEPVH001
Two 25mm x 75mm microscope , chamber EVOS and Micromaster inverted microscopes Each for $173.06
12-563-524
evos™
AMEPVH004
One 100mm petri dish EVOS and Micromaster inverted microscopes Each for $173.06
12-563-525
evos™
AMEPVH005
Two 25cm2 flasks, rectangular or triangular EVOS and Micromaster inverted microscopes Each for $173.06
12-563-526
evos™
AMEPVH006
One 75cm2 flask EVOS and Micromaster inverted microscopes Each for $173.06
12-563-496
evos™
AMEPVH011
One 66mm square tray EVOS and Micromaster inverted microscopes Each for $200.40
12-563-497
evos™
AMEPVH013
Four 35mm petri dishes (EU) EVOS and Micromaster inverted microscopes Each for $200.40
12-563-498
evos™
AMEPVH014
Two 50mm petri dishes (EU) EVOS and Micromaster inverted microscopes Each for $200.40
12-563-493
evos™
AMEPVH008
One 75cm2 BD/Greiner T-75 flask EVOS and Micromaster inverted microscopes Each for $173.06
12-563-495
evos™
AMEPVH010
One 25cm2 BD/Greiner T-25 flask EVOS and Micromaster inverted microscopes Each for $173.06
12-563-494
evos™
AMEPVH009
All vessel types (universal) EVOS and Micromaster inverted microscopes Each for $217.77
12-563-522
evos™
AMEPVH002
Four 35mm petri dishes EVOS and Micromaster inverted microscopes Each for $173.06
12-563-527
evos™
AMEPVH007
One standard hemacytometer EVOS and Micromaster inverted microscopes Each for $173.06
Description & Specifications
Specifications
Product Type Vessel Holder
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.