EVOS™ EVOS™ Onstage Incubator Gas Line
The EVOS™ Onstage Incubator Gas Line connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS™ Onstage Incubator and helps precisely control the CO2 and O2 levels
Supplier: EVOS™ AMEP4732
Description
When performing live cell imaging studies, it is essential that the environmental conditions are precisely controlled. The EVOS™ Onstage Incubator has three gas line ports, enabling studies of cells under normal, as well as hypoxic, conditions.Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Onstage Incubator Gas Line | |
EVOS | |
EVOS™ FL Auto Imaging System | |
1 each |
For Research Use Only. Not for use in diagnostic procedures.