Please login to your online account to display your discounted pricing

EVOS™ Imaging System Accessory, Microscope Objectives


Components manufactured to ensure publication-quality images

Manufacturer: evos™  AMEP4623

 View more versions of this product

Catalog No. 12-563-465

  • / Each
 

Description & Specifications

Specifications

Compatibility EVOS™ FL Auto Imaging System, EVOS™ FL Color Imaging System, EVOS™ FL Imaging System
Quantity 1 Each
Description EVOS™ Objective Lens, 10X, Plan Fluorite, Long Working Distance, 0.30 Numerical Aperture, 8.3mm Working Distance, This Fluorite Objective is Ideal For Fluorescence and Demanding Transmitted Light Applications
Type Objective Lens
Numerical Aperture 0.30
Magnification Power 10X
Product Type EVOS™ LWD fluorite objective
For Use With (Equipment) EVOS™ digital microscope