Please login to your online account to display your discounted pricing

EVOS™ Imaging System Accessory, Microscope Objectives


Components manufactured to ensure publication-quality images

Manufacturer: evos™  AMEP4681

 View more versions of this product

Catalog No. 12-563-536

  • / Each
 

Description & Specifications

Specifications

Magnification Power 10X
For Use With (Equipment) EVOS™ digital microscope
Numerical Aperture 0.25
Product Type EVOS™ LWD fluorite phase contrast objective