missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EVOS™ EVOS™ 4X Objective, fluorite, LWD

Components manufactured to ensure publication-quality images

$4,736.98 - $4,736.98

Specifications

For Use With (Equipment) EVOS™ digital microscope
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Magnification Power Numerical Aperture Price Quantity  
Catalog Number Mfr. No. Product Type Magnification Power Numerical Aperture Price Quantity  
12-563-351
View Documents
EVOS™
AMEP4700
EVOS™ coverslip-corrected oil objective 100X 1.28 Each for $4,736.98
 
Description

Description

RMS-threaded objectives with 45mm parfocal distance

Specifications

Specifications

EVOS™ digital microscope