Please login to your online account to display your discounted pricing

EVOS™ Imaging System Accessory, Microscope Objectives


Components manufactured to ensure publication-quality images

Manufacturer: evos™  AMEP4700*

 View more versions of this product

Catalog No. 12-563-351

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) EVOS™ digital microscope
Magnification Power 100X
Product Type EVOS™ coverslip-corrected oil objective
Immersion Type Oil
Numerical Aperture 1.28