Please login to your online account to display your discounted pricing

EVOS™ Imaging System Accessory, Microscope Objectives


Components manufactured to ensure publication-quality images

Manufacturer: evos™  AMEP4682

 View more versions of this product

Catalog No. 12-563-537

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) EVOS™ digital microscope
Magnification Power 20X
Product Type EVOS™ LWD fluorite phase contrast objective
Numerical Aperture 0.40