Please login to your online account to display your discounted pricing

EVOS™ Imaging System Accessory, Microscope Objectives


Components manufactured to ensure publication-quality images

Manufacturer: evos™  AMEP4634

 View more versions of this product

Catalog No. 12-563-457

  • / Each
 

Description & Specifications

Specifications

Magnification Power 20X
Numerical Aperture 0.40
Product Type EVOS™ LWD achromatic phase contrast objective
For Use With (Equipment) EVOS™ digital microscope