Invitrogen™ EVOS™ Light Cube, Qdot™ 605
The revolutionary design of the EVOS™ light cubes combines a high intensity, long lifetime light source (LED), condensing optics, dichroic mirror and filters into a compact, adjustable, easy-to-install accessory for fluorescence detection on the EVOS™ imaging systems and the Countess II FL.
$8,770.40 - $8,770.40
Specifications
For Use With (Equipment) | EVOS™ FL Imaging System, EVOS™ FL Auto 1 and 2 Imaging System, EVOS™ M5000/M7000 Imaging System, Countess™ II FL Automated Cell Counter |
---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
12-563-485
|
Invitrogen™
AMEP4666 |
Q-Dot 525-800 Fluorescent light cube |
Each for $8,770.40
|
|
Description
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
EVOS™ FL Imaging System, EVOS™ FL Auto 1 and 2 Imaging System, EVOS™ M5000/M7000 Imaging System, Countess™ II FL Automated Cell Counter |