Invitrogen™ EVOS™ Light Cube, Qdot™ 605
The revolutionary design of the EVOS™ light cubes combines a high intensity, long lifetime light source (LED), condensing optics, dichroic mirror and filters into a compact, adjustable, easy-to-install accessory for fluorescence detection on the EVOS™ imaging systems and the Countess II FL.
Manufacturer:  Invitrogen™ AMEP4666
 View more versions of this product
Catalog No. 12-563-485
Description
Description
  • Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
Specifications
EVOS™ FL Auto Imaging System, EVOS™ FL Color Imaging System, EVOS™ FL Imaging System
1 Each
EVOS™ FL Imaging System, EVOS™ FL Auto 1 and 2 Imaging System, EVOS™ M5000/M7000 Imaging System, Countess™ II FL Automated Cell Counter
EVOS™ Qdot™ WB 525-800 Light Cube, LED, Unique, For Use With EVOS™ FL Auto Imaging System, EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, Fluorophores with Excitation and Emission Maxima Near 445 and 525 to 800nm
LED Light Cube
Q-Dot 525-800 Fluorescent light cube