Please login to your online account to display your discounted pricing
EVOS™ Digital Color Fluorescence Microscope
All-in-one digital inverted microscope produces extraordinary on-screen images
Products
Catalog Number Mfr. No. Price Quantity    
12-563-340
invitrogen™
AMEFC4300US
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Unique LED light cube system. Light engine outputs remarkable intensity over a short light path to deliver incomparable excitation. EVOS Light Cubes are easily installed and maintenance-free. Digitally controlled LED light sources allow precise adjustment of illumination levels, dramatically improving control over photobleaching.
  • LED light cubes last >50,000 hours: no need to change bulbs
  • Instant-on; no warm up or cool down required
  • Precision optics for excellent image quality
  • Brightfield and phase contrast
  • Advanced ergonomic design: microscopy made comfortable
  • Small footprint; fits inside cell culture hoods
  • Integrated computer and imaging software
  • High-sensitivity digital camera
  • High-resolution, 15 in. LCD
  • Single switch turns on entire workstation
  • Streamlined software and user interface
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.