missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Caplugs™ (Evergreen Scientific) Transport Vials

Manufacturer:  Caplugs™ (Evergreen Scientific) 2403007060

 View more versions of this product

Catalog No. 05-552-6



Specifications

Specifications

CE, ISO 9001:2008
17 x 54mm
Cap
1000 Case
0.23 oz.
Non-sterile
0.66 x 2.12 in.
Collecting or transporting general purpose specimens
With Cap
Transport Vial
Polypropylene
7 mL
Documents