Caplugs™ (Evergreen Scientific) Test Tubes-R-Us™ General Purpose Polystyrene Test Tubes
Precision molded from premium-grade plastics—ideal for routine laboratory analyses and procedures
Manufacturer:  Caplugs™ (Evergreen Scientific) 2142012010
 View more versions of this product
Catalog No. 05-551-9
Description
Description
  • For use with high-volume clinical chemistry analzyers
  • Polystyrene tubes provide clarity and are appropriate for use with mild acidic (except oxidizing acids) or alkali solutions, alcohols (except higher alcohols) and salt
  • Also available are polypropylene tubes (see cat. no. 22-047-416), which are more chemically resistant and are appropriate for use with organic solvents and stronger acids (except oxidizing acids) and alkalis, alcohols, chlorinated and aromatic hydrocarbons (except xylene) and ketones
  • Cat. No. 05-558-32E can also be used with Beckman™ and Tecan™ analyzers
  • Tubes are bulk packed, non-sterile, with or without caps, round or conical bottom
  • Stopper caps and sterile tubes are also available
Specifications
Specifications
12mm
5mL
Without Cap
Polystyrene
Round Bottom
Polystyrene tubes provide clarity and are appropriate for use with mild acidic (except oxidizing acids) or alkali solutions, alcohols (except higher alcohols) and salt
5mL
75mm
General Purpose Test Tube
Non-sterile
Documents