Caplugs™ Sera-Separa™ Serum Filters
Manufacturer:  Caplugs™ 2083193020
 View more versions of this product
Catalog No. 05-551-5
Description
Description
  • Flange is inserted into evacuated blood tube and pushed downward
  • Wide body style prevents sample probes from hitting side walls
  • Also for use in removing fibrin and macro particles from serum to prevent line clogging
  • Can be used with Evergreen serum tubes
Specifications
Specifications
0.16 oz., 5mL
2.62 in.
1000 Pack
5mL
EverGreen Serum tubes
66.5mm
0.16 oz.
Serum Filter
Documents