Please login to your online account to display your discounted pricing

Polyethylene Stopper Caps


Fit test tubes, culture tubes and sample cups for liquid handling

Manufacturer: caplugs  3001006020

 View more versions of this product

Catalog No. 02-544-73A

  • / Pack of 1000 Each
 

Description & Specifications

Specifications

Material Polyethylene
Product Type Flat Top Cap
Color Natural
For Use With (Equipment) Automated analyzer sample cups