missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Evans Blue, ACROS Organics

$39.45 - $39.45

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.793
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-UHFFFAOYSA-J
Synonym C.I. 23860
PubChem CID 24832074
IUPAC Name tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)C)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC195550050 SDS View Documents Acros Organics
AC195550050
5g Glass bottle Each for $39.45
 
AC195550250 SDS View Documents Acros Organics
AC195550250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

314-13-6
960.793
KBNIFDASRCWYGC-UHFFFAOYSA-J
24832074
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)C)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]
C34H24N6Na4O14S4
MFCD00004021
C.I. 23860
tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate

Specifications

314-13-6
MFCD00004021
KBNIFDASRCWYGC-UHFFFAOYSA-J
tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
24832074
Glass bottle
5g
C34H24N6Na4O14S4
C.I. 23860
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)C)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]
960.793
960.78
Solubility in water: 280g/L (20°C). Other solubilities: soluble in alcohol, acids, alkalies, insoluble in ether
Evans Blue