Please login to your online account to display your discounted pricing

EVA1 Mouse anti-Human, Mouse, PE, Clone: G9P3-1, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen EVA1
Clone G9P3-1
Host Species Murine
Gene Alias Epithelial V-like antigen 1, Myelin protein zero-like 2
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122453

 
life technologies
LS12974280
25μg Each for $158.05

501122454

 
life technologies
LS12974282
100μg Each for $477.05
Description & Specifications

Specifications

Antigen EVA1
Clone G9P3-1
Host Species Murine
Gene Alias Epithelial V-like antigen 1, Myelin protein zero-like 2
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody G9P3-1 reacts with human and mouse epithelial V-like antigen 1 (EVA1), also known as myelin protein zero-like 2 (MPZL2). EVA1 is a low affinity adhesion type I transmembrane glycoprotein that bears an immunolobulin V-type domain. This glycoprotein is expressed on some epithelial tissues and also on immature thymocytes in the DN3 stage. The DN3 thymocytes are characterized by CD4 and CD8 double negative cells that are CD25 positive and CD44 negative. EVA1 has a role in immature thymocyte development, and thymic regeneration.