missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium rod, 9.52 mm (0.375 in.) dia., 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 44545HF

 View more versions of this product

Catalog No. AA44545HF



Description

Description

  • Packaged in mineral oil
Specifications

Specifications

Europium rod
10 mm
7440-53-1
Eu
UN3208
OGPBJKLSAFTDLK-UHFFFAOYSA-N
europium
151.964
CHEBI:32999
99.9%
≈0.37g/mm
≈9.52 mm (0.375 in.) dia.
(metals basis)
MFCD00010992
acmc-1bj8v
[Eu]
Odorless
23981
≈0.37g/mm,Cast Rod,diameter approximate
Gray-Silver

Chemical Identifiers

7440-53-1
151.964
OGPBJKLSAFTDLK-UHFFFAOYSA-N
23981
europium
Eu
MFCD00010992
acmc-1bj8v
CHEBI:32999
[Eu]
Safety and Handling

Safety and Handling

GHS H Statement
H250-H261

P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H250-H261-H500

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-161-7

TSCA : Yes

storageNote1 :

  • Packaged in mineral oil

Recommended Storage : Ambient temperatures

Documents