missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium pieces, sublimed dendritic, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1239406*

 View more versions of this product

Catalog No. AA1239406



Description

Description

  • Packaged under mineral oil
Specifications

Specifications

Europium pieces
Gray-Silver
Sublimed dendritic
Odorless
Eu
UN3208
OGPBJKLSAFTDLK-UHFFFAOYSA-N
europium
23981
Pieces
Air & Moisture Sensitive
5g
7440-53-1
(REO)
MFCD00010992
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
[Eu]
151.964
CHEBI:32999
99.9%

Chemical Identifiers

7440-53-1
151.964
OGPBJKLSAFTDLK-UHFFFAOYSA-N
23981
europium
Eu
MFCD00010992
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
CHEBI:32999
[Eu]
Safety and Handling

Safety and Handling

GHS H Statement
H250-H261

GHS P Statement
P210-P222-P231+P232-P280-P233-P370+P378b-P335+P334-P422a-P402+P404-P501a

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under mineral oil

Documents