Learn More
Europium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01239403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under mineral oil
Specifications
Gray-Silver | |
Sublimed dendritic | |
(REO) | |
MFCD00010992 | |
1 g | |
OGPBJKLSAFTDLK-UHFFFAOYSA-N | |
europium | |
23981 | |
99.9% | |
Air and moisture sensitive |
Europium pieces | |
7440-53-1 | |
Eu | |
UN3208 | |
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil | |
[Eu] | |
151.96 | |
CHEBI:32999 | |
Odorless | |
Pieces |
Chemical Identifiers
7440-53-1 | |
151.96 | |
OGPBJKLSAFTDLK-UHFFFAOYSA-N | |
23981 | |
europium |
Eu | |
MFCD00010992 | |
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil | |
CHEBI:32999 | |
[Eu] |
Safety and Handling
GHS H Statement
H250-H261
P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H250-H261-H500
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber : 231-161-7
TSCA : Yes
storageNote1 :
- Packaged under mineral oil
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only