Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) trifluoromethanesulfonate, 98%

Chemical Identifiers

Chemical Name or Material Europium(III) trifluoromethanesulfonate
CAS 52093-25-1
Linear Formula Eu(OSO2CF3)3
Formula Weight 599.15
MDL Number MFCD00209587
Synonym Europium(III) triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4030509

 
alfa aesar™
4030509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 599.15
Chemical Name or Material Europium(III) trifluoromethanesulfonate
Physical Form Powder
Assay 98%
Sensitivity Hygroscopic
Linear Formula Eu(OSO2CF3)3
MDL Number MFCD00209587
CAS 52093-25-1
Synonym Europium(III) triflate
Color White
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.