missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) trifluoroacetate hydrate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4463418

 View more versions of this product

Catalog No. AA4463418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Europium (III) trifluoroacetate hydrate
Eu(OOCCF3)3·xH2O
UN3288
490.97 (Anhydrous)
99.9%
(metals basis)
MFCD05663782
Soluble in most polar and non polar solids.
Crystalline
50g

Chemical Identifiers

Eu(OOCCF3)3·xH2O
MFCD05663782
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H300

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents