missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Europium(III) thenoyltrifluoroacetonate, trihydrate, 95%, Thermo Scientific Chemicals
$482.25 - $482.25
Chemical Identifiers
CAS | 21392-96-1 |
---|---|
Molecular Formula | C24H12EuF9O6S3·3H2O |
Molecular Weight (g/mol) | 869.54 |
MDL Number | MFCD00150912 |
InChI Key | LSMLVRQTTAIDBZ-BSWAEIBTSA-N |
Synonym | tris 4,4,4-trifluoro-1-2-thienyl-1,3-butanediono europium iii hydrate |
PubChem CID | 91873327 |
IUPAC Name | erbium;(Z)-1,1,1-trifluoro-4-hydroxy-4-thiophen-2-ylbut-3-en-2-one;hydrate |
SMILES | C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.O.[Er] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423190010
|
Thermo Scientific Chemicals
423190010 |
1 g | Glass bottle |
Each for $482.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21392-96-1 | |
869.54 | |
LSMLVRQTTAIDBZ-BSWAEIBTSA-N | |
91873327 | |
C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.O.[Er] |
C24H12EuF9O6S3·3H2O | |
MFCD00150912 | |
tris 4,4,4-trifluoro-1-2-thienyl-1,3-butanediono europium iii hydrate | |
erbium;(Z)-1,1,1-trifluoro-4-hydroxy-4-thiophen-2-ylbut-3-en-2-one;hydrate |
Specifications
21392-96-1 | |
Yellow | |
Authentic | |
Glass bottle | |
MFCD00150912 | |
tris 4,4,4-trifluoro-1-2-thienyl-1,3-butanediono europium iii hydrate | |
LSMLVRQTTAIDBZ-BSWAEIBTSA-N | |
erbium;(Z)-1,1,1-trifluoro-4-hydroxy-4-thiophen-2-ylbut-3-en-2-one;hydrate | |
91873327 | |
95% | |
Europium(III) thenoyltrifluoroacetonate, trihydrate, 95% |
142.0°C to 147.0°C | |
94% min. | |
95% | |
C24H12EuF9O6S3·3H2O | |
1 g | |
Solubility in water: insoluble | |
C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.O.[Er] | |
869.54 | |
869.54 | |
Crystalline Powder or Crystals |
RUO – Research Use Only