missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) oxide, REacton™, nanopowder, 99.999% (REO)

Manufacturer:  Alfa Aesar™ 4405222

 View more versions of this product

Catalog No. AA4405222



Specifications

Specifications

Europium (III) oxide
(REO)
MFCD00010997
dieuropium trioxide
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium(3+);oxygen(2-)
159371
30-50nm APS Powder
Reacton™
Hygroscopic
100g
1308-96-9
Eu2O3
14,3904
Insoluble in water.
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
351.925
351.92
99.999%
30 to 50nm APS
2291°C

Chemical Identifiers

1308-96-9
351.925
AEBZCFFCDTZXHP-UHFFFAOYSA-N
159371
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
Eu2O3
MFCD00010997
dieuropium trioxide
europium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 215-165-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents