missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) oxide, REacton™, 99.996% (REO)

Manufacturer:  Alfa Aesar™ 4434722

 View more versions of this product

Catalog No. AA4434722



Specifications

Specifications

Europium (III) oxide
(REO)
MFCD00010997
dieuropium trioxide
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium(3+);oxygen(2-)
159371
Powder
Reacton™
2291°C
1308-96-9
Eu2O3
14,3904
Insoluble in water.
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
351.925
351.92
99.996%
Hygroscopic
100g

Chemical Identifiers

1308-96-9
351.925
AEBZCFFCDTZXHP-UHFFFAOYSA-N
159371
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
Eu2O3
MFCD00010997
dieuropium trioxide
europium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 215-165-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents