missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium(III) oxide, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer:  Thermo Fisher Scientific 1129903

 View more versions of this product

Catalog No. AA1129903



Specifications

Specifications

Europium(III) oxide
99.99% (REO)
MFCD00010997
dieuropium trioxide
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
351.925
351.92
White
1308-96-9
Eu2O3
White powder or chunks
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium(3+);oxygen(2-)
159371
Powder or Chunks
1g

Chemical Identifiers

1308-96-9
351.925
AEBZCFFCDTZXHP-UHFFFAOYSA-N
159371
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
Eu2O3
MFCD00010997
dieuropium trioxide
europium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 215-165-6

Documents