Learn More
Europium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01130022
Description
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Europium(III) oxide | |
White | |
Powder | |
99.9% (REO) | |
MFCD00010997 | |
Insoluble in water. | |
[O--].[O--].[O--].[Eu+3].[Eu+3] | |
351.93 | |
351.92 | |
White powder or chunks |
2291°C | |
1308-96-9 | |
100 g | |
Eu2O3 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
europium(3+);oxygen(2-) | |
159371 | |
Hygroscopic | |
7.42g/cm3 |
Chemical Identifiers
1308-96-9 | |
351.93 | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
159371 | |
[O--].[O--].[O--].[Eu+3].[Eu+3] |
Eu2O3 | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
europium(3+);oxygen(2-) |
Safety and Handling
EINECSNumber : 215-165-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only