Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) oxalate hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Europium(III) oxalate hydrate
CAS 152864-32-9
Linear Formula Eu2(C2O4)3·xH2O
Formula Weight 567.98 (Anhydrous)
MDL Number MFCD00150143
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1528804

  SDS
alfa aesar™
1528804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1528809

  SDS
alfa aesar™
1528809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 567.98 (Anhydrous)
Linear Formula Eu2(C2O4)3·xH2O
Sensitivity Hygroscopic
Color White
Physical Form Crystalline
Assay Percent Notes (REO)
MDL Number MFCD00150143
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Europium(III) oxalate hydrate
Assay 99.9%
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
TSCA TSCA
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Quantity 2g
CAS 152864-32-9
UN Number UN3288
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.